Assay Details
| Gene Symbol: CHST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358597.4 | XP_003358645.1 | 1010 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003358597.4 transcripts.
Set Membership: |
|

CHST2
Genomic Map