Assay Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127070.5 | XP_003127118.3 | 733 | 81 | ||
XM_021097196.1 | XP_020952855.1 | 733 | 81 | |||
XM_021097197.1 | XP_020952856.1 | 733 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
