Assay Details
| Gene Symbol: RIPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996625.2 | XP_013852079.1 | 877 | 83 | ||
| XM_021089139.1 | XP_020944798.1 | 943 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RIPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996625.2 | XP_013852079.1 | 877 | 83 | ||
| XM_021089139.1 | XP_020944798.1 | 943 | 83 | |||
Set Membership: |
|