Assay Details
| Gene Symbol: CSGALNACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926417.6 | XP_001926452.3 | 1319 | 87 | ||
| XM_021072397.1 | XP_020928056.1 | 1616 | 87 | |||
| XM_021072398.1 | XP_020928057.1 | 1399 | 87 | |||
| XM_021072399.1 | XP_020928058.1 | 1318 | 87 | |||
| XM_021072401.1 | XP_020928060.1 | 1319 | 87 | |||
| XM_021072402.1 | XP_020928061.1 | 1318 | 87 | |||
| XM_021072403.1 | XP_020928062.1 | 1318 | 87 | |||
| XM_021072404.1 | XP_020928063.1 | 1318 | 87 | |||
More Information
Set Membership: |
|

CSGALNACT2
Genomic Map