Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993402.1 | XP_013848856.1 | 1454 | 63 | ||
| XM_021101841.1 | XP_020957500.1 | 1982 | 63 | |||
| XM_021101852.1 | XP_020957511.1 | 1844 | 63 | |||
| XM_021101859.1 | XP_020957518.1 | 1791 | 63 | |||
| XM_021101869.1 | XP_020957528.1 | 1781 | 63 | |||
More Information
Set Membership: |
|

FRMD6
Genomic Map