Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101338.1 | XP_020956997.1 | 1824 | 84 | ||
| XM_021101341.1 | XP_020957000.1 | 1760 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101338.1 | XP_020956997.1 | 1824 | 84 | ||
| XM_021101341.1 | XP_020957000.1 | 1760 | 84 | |||
Set Membership: |
|