Assay Details
| Gene Symbol: SYT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928804.6 | XP_001928839.3 | 1173 | 108 | ||
| XM_013996993.2 | XP_013852447.1 | 1098 | 108 | |||
More Information
Set Membership: |
|

SYT11
Genomic Map