Assay Details
| Gene Symbol: LOC100521938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673242.3 | XP_005673299.1 | 1043 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005673242.3 transcripts.
Set Membership: |
|

LOC100521938
Genomic Map