Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133908.6 | XP_003133956.4 | 2718 | 118 | ||
| XM_005672430.3 | XP_005672487.1 | 2674 | 118 | |||
| XM_021076647.1 | XP_020932306.1 | 2855 | 118 | |||
| XM_021076648.1 | XP_020932307.1 | 2594 | 118 | |||
More Information
Set Membership: |
|

FYB1
Genomic Map