Assay Details
Gene Symbol: LOC100524336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066843.1 | XP_020922502.1 | 998 | 121 | ||
XM_021066844.1 | XP_020922503.1 | 998 | 121 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
