Assay Details
| Gene Symbol: MCOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127906.4 | XP_003127954.1 | 4937 | 69 | ||
| XM_013988836.2 | XP_013844290.1 | 4767 | 69 | |||
| XM_013988837.2 | XP_013844291.1 | 1371 | 69 | |||
| XM_021096340.1 | XP_020951999.1 | 1364 | 69 | |||
More Information
Set Membership: |
|

MCOLN2
Genomic Map