Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985693.2 | XP_013841147.1 | 947 | 73 | ||
| XM_013985694.2 | XP_013841148.1 | 1401 | 73 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013985693.2, XM_013985694.2 transcripts.
Set Membership: |
|

GPR85
Genomic Map