Assay Details
| Gene Symbol: MAPK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091323.1 | XP_020946982.1 | 717 | 63 | ||
| XM_021091324.1 | XP_020946983.1 | 716 | 63 | |||
| XM_021091325.1 | XP_020946984.1 | 812 | 63 | |||
| XM_021091326.1 | XP_020946985.1 | 734 | 63 | |||
| XM_021091327.1 | XP_020946986.1 | 734 | 63 | |||
| XM_021091328.1 | XP_020946987.1 | 1630 | 63 | |||
| XM_021091329.1 | XP_020946988.1 | 1629 | 63 | |||
More Information
Set Membership: |
|

MAPK11
Genomic Map