Assay Details
| Gene Symbol: TMEM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097051.1 | XP_020952710.1 | 1348 | 108 | ||
| XM_021097060.1 | XP_020952719.1 | 1357 | 108 | |||
| XM_021097069.1 | XP_020952728.1 | 846 | 108 | |||
| XM_021097078.1 | XP_020952737.1 | 687 | 108 | |||
More Information
Set Membership: |
|

TMEM62
Genomic Map