Assay Details
| Gene Symbol: VANGL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663204.3 | XP_005663261.2 | 5103 | 127 | ||
| XM_021089572.1 | XP_020945231.1 | 5137 | 127 | |||
More Information
Set Membership: |
|

VANGL2
Genomic Map