Assay Details
| Gene Symbol: PREB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125297.4 | XP_003125345.1 | 1227 | 117 | ||
| GenBank mRNA | AK236497.1 | - | 7 - 8 | 1200 | 117 | |
| AK389278.1 | - | 7 - 8 | 1202 | 117 | ||
More Information
Set Membership: |
|

PREB
Genomic Map