Assay Details
| Gene Symbol: LOC110259329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083607.1 | XP_020939266.1 | 102 | 83 | ||
| GenBank mRNA | AK230568.1 | - | 1 - 2 | 63 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021083607.1 transcripts.
Set Membership: |
|

LOC110259329
Genomic Map