Assay Details
| Gene Symbol: LOC110259329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083606.1 | XP_020939265.1 | 815 | 95 | ||
| XM_021083607.1 | XP_020939266.1 | 754 | 95 | |||
| XM_021083608.1 | XP_020939267.1 | 761 | 95 | |||
| XM_021083609.1 | XP_020939268.1 | 821 | 95 | |||
| GenBank mRNA | AK230568.1 | - | 6 - 7 | 715 | 95 | |
More Information
Set Membership: |
|

LOC110259329
Genomic Map