Assay Details
| Gene Symbol: U2AF1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127055.4 | XP_003127103.1 | 325 | 109 | ||
| XM_013998342.2 | XP_013853796.1 | 405 | 109 | |||
| XM_013998343.2 | XP_013853797.1 | 342 | 109 | |||
| XM_013998346.2 | XP_013853800.1 | 392 | 109 | |||
| XM_013998347.2 | XP_013853801.1 | 375 | 109 | |||
| XM_013998348.2 | XP_013853802.1 | 371 | 109 | |||
| XM_021097217.1 | XP_020952876.1 | 342 | 109 | |||
| XM_021097218.1 | XP_020952877.1 | 342 | 109 | |||
| XM_021097219.1 | XP_020952878.1 | 348 | 109 | |||
| GenBank mRNA | AK344946.1 | - | 4 - 5 | 362 | 109 | |
More Information
Set Membership: |
|

U2AF1L4
Genomic Map