Assay Details
Gene Symbol: WFIKKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086907.1 | XP_020942566.1 | 1583 | 110 | ||
GenBank mRNA | AK395584.1 | - | 1 - 2 | 380 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
