Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097415.1 | XP_020953074.1 | 786 | 109 | ||
| XM_021097416.1 | XP_020953075.1 | 698 | 109 | |||
| GenBank mRNA | AK232198.1 | - | 4 - 5 | 685 | 109 | |
| AK346670.1 | - | 5 - 6 | 666 | 109 | ||
| AK397453.1 | - | 4 - 5 | 697 | 109 | ||
More Information
Set Membership: |
|

CHMP2A
Genomic Map