Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067093.1 | XP_020922752.1 | 5659 | 92 | ||
XM_021067094.1 | XP_020922753.1 | 5648 | 92 | |||
XM_021067095.1 | XP_020922754.1 | 5644 | 92 | |||
XM_021067096.1 | XP_020922755.1 | 5626 | 92 | |||
XM_021067097.1 | XP_020922756.1 | 5569 | 92 | |||
XM_021067098.1 | XP_020922757.1 | 5558 | 92 | |||
XM_021067099.1 | XP_020922758.1 | 5536 | 92 | |||
XM_021067100.1 | XP_020922759.1 | 5525 | 92 | |||
GenBank mRNA | AK240542.1 | - | 1 | 717 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
