Assay Details
| Gene Symbol: GTF2H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994506.2 | XP_013849960.2 | 2929 | 123 | ||
| GenBank mRNA | AK343930.1 | - | 15 | 2618 | 123 | |
More Information
Set Membership: |
|

GTF2H1
Genomic Map