Assay Details
| Gene Symbol: BAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082883.1 | XP_020938542.1 | 862 | 85 | ||
| GenBank mRNA | AK231268.1 | - | 4 | 726 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082883.1 | XP_020938542.1 | 862 | 85 | ||
| GenBank mRNA | AK231268.1 | - | 4 | 726 | 85 | |
Set Membership: |
|