Assay Details
| Gene Symbol: CHST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093819.1 | XP_020949478.1 | 470 | 65 | ||
| XM_021093820.1 | XP_020949479.1 | 475 | 65 | |||
| GenBank mRNA | AK231312.1 | - | 1 | 120 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021093819.1, XM_021093820.1 transcripts.
Set Membership: |
|

CHST4
Genomic Map