Assay Details
| Gene Symbol: TRAPPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123147.5 | XP_003123195.1 | 663 | 98 | ||
| XM_003123149.5 | XP_003123197.1 | 749 | 98 | |||
| GenBank mRNA | AK346355.1 | - | 1 | 624 | 98 | |
More Information
Set Membership: |
|

TRAPPC5
Genomic Map