Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928870.6 | XP_001928905.1 | 1955 | 103 | ||
| XM_003359020.4 | XP_003359068.1 | 2096 | 103 | |||
| GenBank mRNA | AK347753.1 | - | 2 | 2021 | 103 | |
More Information
Set Membership: |
|

NFIL3
Genomic Map