Assay Details
| Gene Symbol: LASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067060.1 | XP_020922719.1 | 2649 | 86 | ||
| XM_021067061.1 | XP_020922720.1 | 2326 | 86 | |||
| GenBank mRNA | AK348462.1 | - | 1 | 314 | 86 | |
More Information
Set Membership: |
|

LASP1
Genomic Map