Assay Details
| Gene Symbol: LOC100523123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091683.1 | XP_020947342.1 | 1927 | 75 | ||
| GenBank mRNA | AK349930.1 | - | 1917 | 75 | ||
| AK397188.1 | - | 9 | 1918 | 75 | ||
| AK397233.1 | - | 1917 | 75 | |||
| AK397397.1 | - | 1917 | 75 | |||
| AK397407.1 | - | 9 | 1917 | 75 | ||
More Information
Set Membership: |
|

LOC100523123
Genomic Map