Assay Details
| Gene Symbol: PPP1R15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656671.3 | XP_005656728.1 | 4341 | 117 | ||
| GenBank mRNA | AK390786.1 | - | 1 | 816 | 117 | |
More Information
Set Membership: |
|

PPP1R15B
Genomic Map