Assay Details
| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661962.3 | XP_005662019.1 | 2045 | 67 | ||
| GenBank mRNA | AK391983.1 | - | 1 | 525 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661962.3 | XP_005662019.1 | 2045 | 67 | ||
| GenBank mRNA | AK391983.1 | - | 1 | 525 | 67 | |
Set Membership: |
|