Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358761.4 | XP_003358809.1 | 2531 | 85 | ||
| XM_005670112.3 | XP_005670169.1 | 2498 | 85 | |||
| XM_005670113.3 | XP_005670170.1 | 2495 | 85 | |||
| XM_005670114.3 | XP_005670171.1 | 2696 | 85 | |||
| XM_005670115.3 | XP_005670172.1 | 2494 | 85 | |||
| XM_005670116.3 | XP_005670173.1 | 2491 | 85 | |||
| XM_005670117.3 | XP_005670174.1 | 2524 | 85 | |||
| XM_021070140.1 | XP_020925799.1 | 2528 | 85 | |||
| GenBank mRNA | AK393123.1 | - | 3 | 2322 | 85 | |
More Information
Set Membership: |
|

CEP19
Genomic Map