Assay Details
| Gene Symbol: NATD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068052.1 | XP_020923711.1 | 2143 | 106 | ||
| XM_021068053.1 | XP_020923712.1 | 2148 | 106 | |||
| GenBank mRNA | AK394078.1 | - | 2 | 1519 | 106 | |
More Information
Set Membership: |
|

NATD1
Genomic Map