Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076260.1 | XP_020931919.1 | 4928 | 110 | ||
| GenBank mRNA | AK397047.1 | - | 1 | 3018 | 109 | |
More Information
Set Membership: |
|

RAB11FIP1
Genomic Map