Assay Details
| Gene Symbol: SWSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005654777.2 | XP_005654834.1 | 829 | 140 | ||
| GenBank mRNA | AK231774.1 | - | 2 | 820 | 140 | |
| AK352412.1 | - | 2 | 823 | 140 | ||
More Information
Set Membership: |
|

SWSAP1
Genomic Map