Assay Details
| Gene Symbol: ZFP36L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013989124.2 | XP_013844578.1 | 1293 | 77 | ||
| XM_021099397.1 | XP_020955056.1 | 1293 | 77 | |||
More Information
Set Membership: |
|

ZFP36L1
Genomic Map