Assay Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128178.4 | XP_003128226.2 | 598 | 128 | ||
| XM_021100183.1 | XP_020955842.1 | 516 | 128 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003128178.4 transcripts.
Set Membership: |
|

SMIM13
Genomic Map