Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243717.1 | NP_001230646.1 | 3 - 4 | 374 | 74 | |
| XM_021063986.1 | XP_020919645.1 | 344 | 74 | |||
| GenBank mRNA | AK347826.1 | - | 1 - 2 | 157 | 74 | |
| CN158733.1 | - | 3 - 4 | 374 | 74 | ||
More Information
Set Membership: |
|

GLRX2
Genomic Map