Assay Details
Gene Symbol: EFCAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243853.1 | NP_001230782.1 | 2 - 3 | 651 | 63 | |
XM_021068405.1 | XP_020924064.1 | 692 | 63 | |||
XM_021068406.1 | XP_020924065.1 | 788 | 63 | |||
GenBank mRNA | AK240134.1 | - | 2 - 3 | 651 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
