Assay Details
Gene Symbol: EFCAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243853.1 | NP_001230782.1 | 4 - 5 | 983 | 94 | |
XM_021068405.1 | XP_020924064.1 | 1024 | 94 | |||
XM_021068406.1 | XP_020924065.1 | 1120 | 94 | |||
GenBank mRNA | AK240134.1 | - | 4 - 5 | 983 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
