Assay Details
Gene Symbol: EFCAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243853.1 | NP_001230782.1 | 5 - 6 | 1184 | 64 | |
XM_021068405.1 | XP_020924064.1 | 1225 | 64 | |||
XM_021068406.1 | XP_020924065.1 | 1321 | 64 | |||
GenBank mRNA | AK240134.1 | - | 5 - 6 | 1184 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
