Assay Details
| Gene Symbol: ARSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244120.1 | NP_001231049.1 | 10 - 11 | 1676 | 82 | |
| XM_021079485.1 | XP_020935144.1 | 1787 | 82 | |||
| XM_021079486.1 | XP_020935145.1 | 1847 | 82 | |||
| GenBank mRNA | AK238509.1 | - | 5 - 6 | 910 | 82 | |
| AK351157.1 | - | 10 - 11 | 1740 | 82 | ||
| AK351168.1 | - | 10 - 11 | 1526 | 82 | ||
| AK351217.1 | - | 10 - 11 | 1654 | 82 | ||
| AK351260.1 | - | 10 - 11 | 1676 | 82 | ||
| AK398154.1 | - | 10 - 11 | 1684 | 82 | ||
More Information
Set Membership: |
|

ARSE
Genomic Map