Assay Details
Gene Symbol: CATSPER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244299.1 | NP_001231228.1 | 5 - 6 | 943 | 84 | |
GenBank mRNA | HQ654521.1 | - | 5 - 6 | 943 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
