Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244827.1 | NP_001231756.1 | 2 - 3 | 136 | 101 | |
| GenBank mRNA | AK399988.1 | - | 2 - 3 | 176 | 101 | |
| EW077775.2 | - | 2 - 3 | 159 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244827.1 transcripts.
Set Membership: |
|

BEX3
Genomic Map