Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244828.1 | NP_001231757.1 | 2 - 3 | 186 | 78 | |
| GenBank mRNA | AK391649.1 | - | 2 - 3 | 153 | 78 | |
| AK399973.1 | - | 2 - 3 | 218 | 78 | ||
| FS643797.1 | - | 2 - 3 | 186 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244828.1 transcripts.
Set Membership: |
|

BEX3
Genomic Map