Assay Details
| Gene Symbol: TREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256777.1 | NP_001243706.1 | 1 - 2 | 154 | 77 | |
| GenBank mRNA | JQ247512.1 | - | 1 - 2 | 154 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001256777.1 transcripts.
Set Membership: |
|

TREM2
Genomic Map