Assay Details
| Gene Symbol: FOSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315740.1 | NP_001302669.1 | 3 - 4 | 513 | 135 | |
| XM_021085708.1 | XP_020941367.1 | 1383 | 135 | |||
| XM_021085709.1 | XP_020941368.1 | 1241 | 135 | |||
| GenBank mRNA | AK349073.1 | - | 3 - 4 | 799 | 135 | |
| KM191330.1 | - | 3 - 4 | 513 | 135 | ||
More Information
Set Membership: |
|

FOSL2
Genomic Map