Assay Details
| Gene Symbol: WARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315752.1 | NP_001302681.1 | 3 - 4 | 426 | 66 | |
| GenBank mRNA | AK238494.1 | - | 3 - 4 | 436 | 66 | |
| AK395995.1 | - | 3 - 4 | 444 | 66 | ||
| EU650788.1 | - | 3 - 4 | 426 | 66 | ||
More Information
Set Membership: |
|

WARS2
Genomic Map