Assay Details
| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315776.1 | NP_001302705.1 | 6 - 7 | 608 | 56 | |
| XM_021085801.1 | XP_020941460.1 | 680 | 56 | |||
| XM_021085802.1 | XP_020941461.1 | 680 | 56 | |||
| GenBank mRNA | JF944892.1 | - | 6 - 7 | 608 | 56 | |
More Information
Set Membership: |
|

RBKS
Genomic Map