Assay Details
| Gene Symbol: CREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099929.2 | NP_001093399.2 | 5 - 6 | 745 | 107 | |
| NM_001361427.1 | NP_001348356.1 | 6 - 7 | 787 | 107 | ||
| XM_021075304.1 | XP_020930963.1 | 668 | 107 | |||
| XM_021075308.1 | XP_020930967.1 | 584 | 107 | |||
| XM_021075309.1 | XP_020930968.1 | 561 | 107 | |||
| XM_021075310.1 | XP_020930969.1 | 436 | 107 | |||
| XM_021075311.1 | XP_020930970.1 | 481 | 107 | |||
| XM_021075312.1 | XP_020930971.1 | 715 | 107 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021075310.1, XM_021075311.1, XM_021075312.1 transcripts.
Set Membership: |
|

CREB1
Genomic Map